Please login to your online account to display your discounted pricing

Talc (Powder/Laboratory), Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  T4500

 View more versions of this product

Catalog No. T4-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Melting Point 800°C
Purity Grade Laboratory Grade
Quantity 500g
Color Off-white
Molecular Formula 3MgO4SiO2H2O
Chemical Name or Material Talc
Identification Pass Test
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
ChemAlert Storage Symbol Gray
Packaging Poly Bottle
MDL Number MFCD00084820
Synonym Soapstone
CAS 14808-60-7
CAS 14807-96-6

Specifications

Chemical Name or Material Talc
CAS 14807-96-6
CAS 14808-60-7
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Molecular Formula 3MgO4SiO2H2O
MDL Number MFCD00084820
Synonym Soapstone
Accessories
 

Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

SDS & Safety Documents