missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Talc, particle size: -200 mesh (<75 μm), Thermo Scientific Chemicals
$98.61 - $865.89
Chemical Identifiers
CAS | 14807-96-6 |
---|---|
Molecular Formula | H2Mg3O12Si4 |
Molecular Weight (g/mol) | 379.26 |
MDL Number | MFCD00792903 |
InChI Key | KNXVOGGZOFOROK-UHFFFAOYSA-N |
Synonym | magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate |
PubChem CID | 16211421 |
IUPAC Name | dioxosilane;oxomagnesium;hydrate |
SMILES | [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC222735000
|
Thermo Scientific Chemicals
222735000 |
500 g | Plastic bottle |
Each for $98.61
|
|
||||
AC222730025
|
Thermo Scientific Chemicals
222730025 |
2.5 kg | Plastic bottle |
Each for $370.64
|
|
||||
AC222730100
|
Thermo Scientific Chemicals
222730100 |
10 kg | Plastic drum |
Each for $865.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14807-96-6 | |
379.26 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
16211421 | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
dioxosilane;oxomagnesium;hydrate |
Specifications
800.0°C | |
14807-96-6 | |
500 g | |
Mg3[Si4O10(OH)2] | |
15,9168 | |
Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkalies | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
379.26 | |
379.28 | |
0.2% (105°C) max. | |
Talc |
White | |
Very Fine Powder | |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
dioxosilane;oxomagnesium;hydrate | |
16211421 | |
Typical chemical properties: SiO2: 59.3%, MgO: 31.5%, Al2O3: 1.9%, Fe2O3: 1.0%, CaO: 0.6% | |
Plastic bottle |
Safety and Handling
EINECSNumber : 238-877-9
RUO – Research Use Only