missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Talc, particle size: -200 mesh (<75 μm), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 222735000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Talc | |
White | |
Very Fine Powder | |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
dioxosilane;oxomagnesium;hydrate | |
16211421 | |
Typical chemical properties: SiO2: 59.3%, MgO: 31.5%, Al2O3: 1.9%, Fe2O3: 1.0%, CaO: 0.6% | |
Plastic bottle |
800.0°C | |
14807-96-6 | |
500 g | |
Mg3[Si4O10(OH)2] | |
15,9168 | |
Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkalies | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
379.26 | |
379.28 | |
0.2% (105°C) max. |
Chemical Identifiers
14807-96-6 | |
379.26 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
16211421 | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
dioxosilane;oxomagnesium;hydrate |
Safety and Handling
EINECSNumber : 238-877-9
RUO – Research Use Only