Please login to your online account to display your discounted pricing

Talc, particle size: -200 mesh (<75μm), ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  222730025

 View more versions of this product

Catalog No. AC222730025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Very Fine Powder
Formula Weight 379.28
Quantity 2.5kg
Color White or almost white
Additional Information residue on 75 um sieve: 0.3% (typically)
Loss on Ignition 6.5% (1050°C) max.
Chemical Name or Material Talc
Composition Typical chemical properties: SiO2: 59.3%, MgO: 31.5%, Al2O3: 1.9%, Fe2O3: 1.0%, CaO: 0.6%
Synonym Magnesium silicate, hydrous
CAS 14807-96-6
Loss on Drying 0.2% (105°C) max.
Packaging Plastic bottle
Molecular Formula H2Mg3O12Si4
Linear Formula Mg3[Si4O10(OH)2]

Specifications

Chemical Name or Material Talc
CAS 14807-96-6
Molecular Formula H2Mg3O12Si4
Linear Formula Mg3[Si4O10(OH)2]
Formula Weight 379.28
Synonym Magnesium silicate, hydrous