Troemner™ Talboys™ Advanced Digital 1000-3 Orbital Shaker
Operates in cold rooms, incubators, and CO2 environments from -10 to 60°C (14 to 140°F). The Troemner™ Talboys™ Advanced Digital 1000-3 Orbital Shaker is ideal for a wide variety of shaking applications.
Manufacturer: Troemner™ 980175
Description
- General purpose shaker
- LED Displays for speed and time
- 3mm orbit
- 5 year industry leading warranty
- Includes a non-skid rubber mat
Specifications
1000-3 | |
3 mm | |
1 second to 160 hours | |
3.6 kg | |
17 x 11 x 4 in. (43.2 x 27.9 x 10.2cm) | |
11.75 x 8.75 in. (29.9 x 22.2cm) |
100-1200 rpm | |
8 lb. | |
TUV | |
Digital | |
120V | |
100-1200 rpm |