Please login to your online account to display your discounted pricing

TAK 715, Tocris™

Potent p38 MAPK inhibitor; anti-inflammatory

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material TAK 715
Assay Percent Range >99%
Molecular Formula C24H21N3OS
Formula Weight 399.51
Synonym N-[4-[2-Ethyl-4-(3-methylphenyl)-5-thiazolyl]-2-pyridinyl]benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

425450

 
r & d systems
RDS425450
50mg Each for $1,050.42

425410

 
r & d systems
RDS425410
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 399.51
Quantity 50mg
Assay Percent Range >99%
CAS 303162-79-0
Chemical Name or Material TAK 715
Molecular Formula C24H21N3OS
Synonym N-[4-[2-Ethyl-4-(3-methylphenyl)-5-thiazolyl]-2-pyridinyl]benzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C