Please login to your online account to display your discounted pricing

TAE Buffer, 10X, Molecular Biology Grade, Calbiochem™, EMD Millipore

A 10X concentrate that can be diluted to a 1X solution containing 40mM Tris, 40mM acetate, and 1mM EDTA

$48.75 - $48.75

Chemical Identifiers

Analyte or Component Names Tris, EDTA, Acetate
Chemical Name or Material TAE Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 40mM Tris, 40mM acetate, and 1mM EDTA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5747971L

 
emd millipore corporation
5747971L
Each for $48.75
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material TAE Buffer
Analyte or Component Names Tris, EDTA, Acetate
DNase None Detected
Synonym Tris-Acetate-EDTA Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 40mM Tris, 40mM acetate, and 1mM EDTA
Solubility Information Easily soluble in cold water
Protease None Detected
Purity Molecular Biology Grade
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.
RNase None Detected
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.