missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tacrine hydrochloride

A potent centrally acting anticholinesterase

$268.32 - $268.32

Chemical Identifiers

CAS 1684-40-8
Molecular Formula C13H15ClN2
Molecular Weight (g/mol) 234.727
MDL Number MFCD00150067
InChI Key ZUFVXZVXEJHHBN-UHFFFAOYSA-N
Synonym tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250
PubChem CID 2723754
IUPAC Name 1,2,3,4-tetrahydroacridin-9-amine;hydrochloride
SMILES C1CCC2=NC3=CC=CC=C3C(=C2C1)N.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6007006 View Documents Alfa Aesar™
J6007006
5g Each for $268.32
 
Description

Description

Solubility
Soluble in aqueous buffers

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1684-40-8
234.727
ZUFVXZVXEJHHBN-UHFFFAOYSA-N
2723754
C1CCC2=NC3=CC=CC=C3C(=C2C1)N.Cl
C13H15ClN2
MFCD00150067
tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250
1,2,3,4-tetrahydroacridin-9-amine;hydrochloride

Specifications

Yellow
5g
C13H15ClN2
UN2811
tacrine hydrochloride, 1,2,3,4-tetrahydroacridin-9-amine hydrochloride, tacrine hcl, hydroaminacrine, tenakrin, 9-amino-1,2,3,4-tetrahydroacridine hydrochloride, tacrine.hcl, unii-4966rng0bu, 1,2,3,4-tetrahydro-9-acridinamine hydrochloride, ccris 7250
ZUFVXZVXEJHHBN-UHFFFAOYSA-N
1,2,3,4-tetrahydroacridin-9-amine;hydrochloride
2723754
Crystalline Solid
Tacrine hydrochloride
283°C to 284°C
1684-40-8
MFCD00150067
14,9024
Soluble in aqueous buffers
C1CCC2=NC3=CC=CC=C3C(=C2C1)N.Cl
234.727
234.73 (Anhydrous)
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : AR9532500

Recommended Storage : 2° to 8°C