missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Tacho Strip for Spinner Turbine

Replacement strip with clear protective foil cover

Supplier:  Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 660060059

Catalog No. 03415512



Description

Description

  • Causes less probe insert damage by better insert sample control
  • Strip with foil protection to improve life span
  • Longer upper barrel stabilizer
    • Easily identified by 3 mm yellow band
  • Limited VT operation possible 40 °C above ambient
  • Can mix with originals during sample changer operation
  • Susceptibility matched to originals
Specifications

Specifications

5 mm Bruker spinner turbines
Cause less probe insert damage
Clear protective foil cover
SDS
Documents

Documents

Promotions

Promotions