Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

$532.67 - $1,628.67

Specifications

Antigen TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB1660P

 
thermo scientific™ lab vision™
RB1660P
Purified Ab with BSA and Azide 1mL Each for $1,508.78

RB1660P0

 
thermo scientific
RB1660P0
Purified Ab with BSA and Azide 100µL Each for $532.67

RB1660P1

 
thermo scientific
RB1660P1
Purified Ab with BSA and Azide 500µL Each for $1,075.35

RB1660P1ABX

 
thermo scientific
RB1660P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $1,191.16

RB1660PABX

 
thermo scientific™ lab vision™
RB1660PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,628.67
Description & Specifications

Specifications

Antigen TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Type Primary
Immunogen A synthetic peptide from the C-terminus of human TACE protein
Isotype IgG

Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

TACE (TNF-α converting enzyme) protein is a membrane bound matalloprotease from ADAM (a disintegrin and metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-α into 17kDA mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGFα, L-selectin and amyloid protein precursor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human TACE protein

Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)

Positive Control: HeLa cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO