Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1660PABX

 View more versions of this product

Catalog No. RB1660PABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide from the C-terminus of human TACE protein
Isotype IgG
Antigen TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1
Format Concentrated
Quantity 1mL
Host Species Rabbit
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Research Discipline Proteases And Inhibitors
Type Primary
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Classification Polyclonal
Item Description 1mL; 1mg/mL; Unlabeled; Purified without BSA and azide

Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

TACE (TNF-α converting enzyme) protein is a membrane bound matalloprotease from ADAM (a disintegrin and metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-α into 17kDA mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGFα, L-selectin and amyloid protein precursor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human TACE protein

Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)

Positive Control: HeLa cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO