Please login to your online account to display your discounted pricing

Thermo Scientific TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific  RB1660P1ABX

 View more versions of this product

Catalog No. RB1660P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 500µL
Host Species Rabbit
Isotype IgG
Format Purified
Cross Reactivity Human
Cross Reactivity Mouse
Applications Western Blotting
Regulatory Status RUO
Antigen TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1
Immunogen A synthetic peptide from the C-terminus of human TACE protein
Clone Polyclonal
Formulation Ab without BSA/Azide (Frozen)
Classification Polyclonal
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline Proteases And Inhibitors

Provide accurate, reproducible results Thermo Scientific™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.

TACE (TNF-α converting enzyme) protein is a membrane bound matalloprotease from ADAM (a disintegrin and metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-α into 17kDA mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGFα, L-selectin and amyloid protein precursor.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of human TACE protein

Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)

Positive Control: HeLa cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO