Please login to your online account to display your discounted pricing

TACA, Tocris


GABAA agonist; Also GABA-T substrate and GABA uptake inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS018150

 View more versions of this product

Catalog No. 018150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 101.1
Chemical Name or Material TACA
Assay Percent Range >98%
Molecular Formula C4H7NO2
CAS 38090-53-8
Synonym trans-4-Aminocrotonic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material TACA
Assay Percent Range >98%
Molecular Formula C4H7NO2
Formula Weight 101.1
Synonym trans-4-Aminocrotonic acid
Accessories

Store at Room Temperature