Please login to your online account to display your discounted pricing

TACA, Tocris

GABAA agonist; Also GABA-T substrate and GABA uptake inhibitor

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material TACA
Assay Percent Range >98%
Molecular Formula C4H7NO2
Formula Weight 101.1
Synonym trans-4-Aminocrotonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

018110

 
r & d systems
RDS018110
10mg Each for $146.74

018150

 
r & d systems
RDS018150
50mg Each for $618.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 101.1
Chemical Name or Material TACA
Assay Percent Range >98%
Molecular Formula C4H7NO2
CAS 38090-53-8
Synonym trans-4-Aminocrotonic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature