Thermo Scientific™ T4 DNA Polymerase
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Manufacturer:  Thermo Scientific™ EP0061
 View more versions of this product
Catalog No. FEREP0061
Specifications
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
  • Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
  • Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
Description & Specifications
Specifications
1mL of 5X Reaction Buffer
Blunting of DNA ends: fill-in 5'-overhangs or/and removal of 3'-overhangs (1, 2); Blunting of PCR products with 3'-dA overhangs; Synthesis of label
T4 DNA Polymerase
100U