Please login to your online account to display your discounted pricing

T16Ainh - A01, Tocris™

Ca2+-activated Cl- channel transmembrane protein 16A (TMEM16A) inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material T16Ainh - A01
Assay Percent Range >98%
Molecular Formula C19H20N4O3S2
Formula Weight 416.52
Synonym 2-[(5-Ethyl-1, 6-dihydro-4-methyl-6-oxo-2-pyrimidinyl)thio]-N-[4-(4-methoxyphenyl)-2-thiazolyl]acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453810

 
r & d systems
RDS453810
10mg Each for $218.74

453850

 
r & d systems
RDS453850
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 416.52
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C19H20N4O3S2
CAS 552309-42-9
Synonym 2-[(5-Ethyl-1, 6-dihydro-4-methyl-6-oxo-2-pyrimidinyl)thio]-N-[4-(4-methoxyphenyl)-2-thiazolyl]acetamide
Chemical Name or Material T16Ainh - A01
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C