missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

t-Butyl4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate, ≥95%, Maybridge

Chemical Identifiers

CAS 365413-31-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H24N2O4S
Formula Weight 340.44
MDL Number MFCD06659069
Chemical Name or Material Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC31023DA SDS Fisher Scientific Company
CC31023DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31023CB SDS Fisher Scientific Company
CC31023CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

365413-31-6
100.0
C16H24N2O4S
MFCD06659069
97%
96.5
97%
340.44
Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate

Specifications

340.44
100.0
365413-31-6
Amber Glass Bottle
97%
97%
1g
MFCD06659069
96.5
C16H24N2O4S
Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.