Please login to your online account to display your discounted pricing

t-Butyl4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate, ≥95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31023CB

 View more versions of this product

Catalog No. CC31023CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 340.44
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber Glass Bottle
Molecular Formula C16H24N2O4S
MDL Number MFCD06659069
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 96.5
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Chemical Name or Material Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
CAS 365413-31-6

Specifications

Chemical Name or Material Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
CAS 365413-31-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H24N2O4S
Formula Weight 340.44
MDL Number MFCD06659069
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents