missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

T-bet, PerCP-Cyanine5.5, clone: eBio4B10 (4B10), eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS45582580

 View more versions of this product

Catalog No. 501130726



Description

Description

T-bet, also known as TBX21, is a T-cell specific transcription factor containing a single T-box DNA-binding domain. It controls the expression of the TH1 cytokine, interferon-gamma. Both Th1 and Th17 cells are crucial in immune regulation and autoimmune disease development. T-bet initiates Th1 lineage development from naive Th precursor cells and directs T cell differentiation to Th1 versus Th17. T-bet is cooperating with Stat4 (signal tranducer and activator of transcription 4) in programming chromatin architecture for Th1 gene expression. Genetic variations in T-bet are associated with susceptibility to aspirin-induced asthma and nasal polyps.

Description: The eBio4B10 monoclonal antibody reacts with mouse and human T-bet. T-bet is a Th1-specific T-box transcription factor critical to the development of the Th1 CD4+ T cell lineage. This is known based on the observations that T-Bet deficient mice have impaired Th1 cell development, and that ectopic expression of T-Bet results in development skewed to the Th1 lineage. T-Bet expression is induced by the Th1 cytokine IFN gamma, and T-Bet also regulates the expression of IFN gamma, likely, at least in part, through the modification of DNA accessibility and histone remodeling. In addition to IFN gamma, T-Bet is also known to regulate the expression of IL-12R beta and IL-2. Moreover, T-Bet plays a role in class-switch recombination in B-cells.Applications Reported: This eBio4B10 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This eBio4B10 antibody has been tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. This antibody can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.For intracellular staining with the eBio4B10 monoclonal antibody, it is recommended to use the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol. Please refer to Best Protocols; Protocol B: One step protocol for intracellular (nuclear) proteins).Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

T-bet
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9UL17, Q9JKD8
T-PET, T-bet, TBET, TBLYM, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, T-cell-specific T-box transcription factor T-bet, transcription factor TBLYM
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Murine, Rhesus Monkey
Flow Cytometry
eBio4B10 (4B10), 4-B10)
PerCP-Cyanine5.5
TBX21
Liquid
TBX21
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
30009, 57765, 694044
Documents

For Research Use Only.