Please login to your online account to display your discounted pricing

T 0156 hydrochloride, Tocris

Highly potent, selective PDE5 inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material T 0156 hydrochloride
Assay Percent Range >99%
Molecular Formula C31H29N5O7·HCl
Formula Weight 620.06
Synonym 1,2-Dihydro-2-[(2-methyl-4-pyridinyl)methyl]-1-oxo-8-(2-pyrimidinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-2,7-naphthyridine-3-carboxylic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

167610

 
r & d systems
167610
10mg Each for $239.63

167650

 
r & d systems
167650
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 620.06
Quantity 10mg
Chemical Name or Material T 0156 hydrochloride
Assay Percent Range >99%
Molecular Formula C31H29N5O7·HCl
CAS 324572-93-2
Synonym 1,2-Dihydro-2-[(2-methyl-4-pyridinyl)methyl]-1-oxo-8-(2-pyrimidinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-2,7-naphthyridine-3-carboxylic acid methyl ester hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C