Please login to your online account to display your discounted pricing

T 0070907, Tocris

Highly potent and selective PPARγ antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material T 0070907
Assay Percent Range >99%
Molecular Formula C12H8N3O3Cl
Formula Weight 277.67
Synonym 2-Chloro-5-nitro-N-4-pyridinylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

230150

 
r & d systems
RDS230150
50mg Each for $794.96

230110

 
r & d systems
RDS230110
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 277.67
Quantity 50mg
Chemical Name or Material T 0070907
Assay Percent Range >99%
Molecular Formula C12H8N3O3Cl
CAS 313516-66-4
Synonym 2-Chloro-5-nitro-N-4-pyridinylbenzamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature