Please login to your online account to display your discounted pricing

Syringic acid, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  132890100

 View more versions of this product

Catalog No. AC132890100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 198.17
Color Grayish-beige to light brown
Quantity 10g
Beilstein 10, 480
Assay 96.0 to 104.0%
Melting Point 206° to 212°C
Assay Percent Range 97%
MDL Number MFCD00002552
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
Chemical Name or Material Syringic acid
Synonym 4-Hydroxy-3, 5-dimethoxybenzoic acid
Packaging Glass bottle
Molecular Formula C9H10O5
CAS 530-57-4
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Syringic acid
CAS 530-57-4
Assay 96.0 to 104.0%
Assay Percent Range 97%
Molecular Formula C9H10O5
Formula Weight 198.17
MDL Number MFCD00002552
Synonym 4-Hydroxy-3, 5-dimethoxybenzoic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents