missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Syrian Hamster IgG Isotype Control, Brilliant Violet™ 711, eBioscience™, Invitrogen™
Syrian Hamster Polyclonal Antibody
Supplier: Fisher Chemical 407491481
Description
Syrian Hamster IgG Polyclonal antibody specifically detects Syrian Hamster IgG in Not Applicable samples. It is validated for Control, Flow CytometrySpecifications
Syrian Hamster IgG | |
Polyclonal | |
Brilliant Violet 711 | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Affinity chromatography | |
RUO | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Syrian Hamster | |
50 μg | |
Not Applicable | |
Isotype Control | |
IgG |