Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Synuclein Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1572P1ABX

 View more versions of this product

Catalog No. MS1572P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Synuclein Ab-2
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone syn211
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Human recombinant alpha-synuclein
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.

Host Species: Mouse

Clone: syn211

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant α-synuclein

Molecular Weight: ∽14kDa

Positive Control: Brain

Cellular Localization: Neuronal cytosol and presynaptic terminals

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO