Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Synuclein Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1572PABX

 View more versions of this product

Catalog No. MS1572PABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human recombinant alpha-synuclein
Quantity 200µL
Clone syn211
Antigen Synuclein Ab-2
Host Species Mouse
Isotype IgG1
Format Concentrated
Cross Reactivity Human
Regulatory Status RUO
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Neuroscience
Conjugate Unlabeled
Classification Monoclonal
Concentration 1mg/mL
Type Primary

Provide accurate, reproducible results Thermo Scientific™ Synuclein Ab-2, Mouse Monoclonal Antibody.

Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.

Host Species: Mouse

Clone: syn211

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Human recombinant α-synuclein

Molecular Weight: ∽14kDa

Positive Control: Brain

Cellular Localization: Neuronal cytosol and presynaptic terminals

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO