Kemtech America™ Synthware™ Vacuum/Inert Gas Adapters with T-bore Glass or Teflon™ Stopcock
Allows vessels with standard taper outer joint to be purged with an inert gas
$101.34 - $140.23
Specifications
Product Type Vacuum/Inert gas adapter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Standard Taper Joint Price Quantity  
Catalog Number Mfr. No. Material Standard Taper Joint Price Quantity  
31500574 Kemtech America™
A881420G
Stopcock: Glass Each for $101.34
31500575 Kemtech America™
A882440G
Stopcock: Glass Each for $106.51
31500576 Kemtech America™
A881420
Stopcock: PTFE Resin Each for $135.14
31500577 Kemtech America™
A882440
Stopcock: PTFE Resin Each for $140.23
Specifications
Description
  • With 4mm glass or Teflon stopcock bore
  • Glass stopcocks have constant pressure retention device
Description & Specifications
Specifications
Vacuum/Inert gas adapter