missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Vacuum Filtration Adapters
Used with plain stem Buchner funnels and pluro-stopper
$68.24 - $97.50
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Product Type | Vacuum filtration adapter |
Catalog Number | Mfr. No. | Standard Taper Joint No. | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint No. | For Use With (Equipment) | Price | Quantity | ||||
31500694
|
Synthware™
A841420 |
14/20 Bottom | Fits Funnel Capacity (mL): 2 to 30 |
Each for $68.24
|
|
||||
31500695
|
Synthware™
A842415 |
24/40 Bottom | Fits Funnel Capacity (mL): 15 to 150 |
Each for $95.94
|
|
||||
31500696
|
Synthware™
A842440 |
24/40 Bottom | Fits Funnel Capacity (mL): 350 to 3000 |
Each for $97.50
|
|
||||
Description
- Hose connection has an O.D. of 10mm at the largest serration
Specifications
Borosilicate Glass (3.3) | |
Vacuum filtration adapter |