Synthware™ Two Neck Pear Shaped Flasks
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Supplier: Synthware™ F341110
Description
- Standard taper inner joint at each end
- Available with either coarse or medium porosity fritted disc
- With two 2 mm glass stopcocks having constant pressure retention devices
Specifications
Two neck pear shaped flask | |
10 mL | |
Center: 14/20; Side: 14/20 |