Synthware™ Storage Tubes with High Vacuum Valve
For long term deoxygenated solvent storage
$131.44 - $225.06
Catalog Number | Mfr. No. | Description | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity | |||
31502239
|
Synthware™
F580820 |
Valve size: 0-8 mm | 19 mm | 30 mm |
Each for $149.97
|
|
|||
31502237
|
Synthware™
F580810 |
Valve size: 0-8 mm | 19 mm | 65 mm |
Each for $149.97
|
|
|||
31502236
|
Synthware™
F580010 |
Valve size: 0-4 mm | 22 mm | 55 mm |
Each for $131.44
|
|
|||
31502238
|
Synthware™
F580020 |
Valve size: 0-4 mm | 22 mm | 110 mm |
Each for $131.44
|
|
|||
31501335
|
Synthware™
F580025 |
Valve Size: 0-4mm | 26 mm | 110 mm |
Each for $156.82
|
|
|||
31501336
|
Synthware™
F580825 |
Valve Size: 0-8mm | 26 mm | 110 mm |
Each for $179.18
|
|
|||
31501337
|
Synthware™
F580050 |
Valve Size: 0-4mm | 28 mm | 120 mm |
Each for $183.47
|
|
|||
31501338
|
Synthware™
F580850 |
Valve Size: 0-8mm | 28 mm | 120 mm |
Each for $208.24
|
|
|||
31501339
|
Synthware™
F580100 |
Valve Size: 0-4mm | 40 mm | 165 mm |
Each for $193.21
|
|
|||
31501340
|
Synthware™
F588100 |
Valve Size: 0-8mm | 40 mm | 165 mm |
Each for $215.09
|
|
|||
31501341
|
Synthware™
F588250 |
Valve Size: 0-8mm | 60 mm | 165 mm |
Each for $225.06
|
|
|||
Description
- High vacuum valve with PTFE O-ring at tip and PTFE wiper (“umbrella”) to protect Viton O-ring on shaft