Synthware™ Single Neck Round Bottom Flasks, Full Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Supplier: Synthware™ F304300
Description
SYNTHWARE round bottom boiling flasks with an extended neck and full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.Specifications
Clear | |
300 mL | |
Organic Synthesis |
Single neck, round bottom flask | |
Borosilicate Glass (3.3) | |
24/40 |