Synthware™ Separatory Funnels with PTFE Stopcock and Stopper
Includes PTFE Stopper
Supplier:  Synthware™ F459025
 View more versions of this product
Catalog No. 31501616
Description
Description
  • Upper outer and lower inner standard taper joints are same size
Specifications
Specifications
Separatory funnel
Glass
2mm
25 mL
19/22
PTFE