Synthware™ Round Bottom Flask with Threaded Inlet
Round bottom, single short neck flask with a standard taper outer joint
Supplier:  Synthware™ F144250
 View more versions of this product
Catalog No. 31501148
Description
Description
  • Threaded side port for insertion of thermometer or bleed tube having O.D. between 5 mm and 7.5 mm
  • Heavy wall design
  • All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Joint: 24/40; With threaded inlet
250 mL