Synthware™ Round Bottom Flask with Threaded Inlet
Round bottom, single short neck flask with a standard taper outer joint
Supplier:  Synthware™ F141425
 View more versions of this product
Catalog No. 31501142
Description
Description
  • Threaded side port for insertion of thermometer or bleed tube having O.D. between 5 mm and 7.5 mm
  • Heavy wall design
  • All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Joint: 14/20; With threaded inlet
25 mL