Synthware™ Recovery Flasks
Used as an evaporation flask in rotary evaporators
Supplier: Synthware™ F314100
Description
- WIth standard taper outer joint
- Heavy wall design for increased safety
- All sizes are hand blown from tubing to ensure uniform wall thickness
Specifications
Recovery flask | |
100 mL | |
24/40 |