Synthware™ Pressure Equalizing Funnels with High Vacuum Valve
With extended tip valve for semi-needle valve adjustment
Supplier: Synthware™ F641425
Description
- With pressure equalizing arm and high vacuum valve
- Top outer and lower inner standard taper joints of same size
Specifications
Valve orifice: 0-4mm | |
25 mL | |
14/20 | |
Pressure Equalizing Funnel with High Vacuum Valve |