Synthware™ Kuderna-Danish Flasks
Shaped to provide continuous flow of heavier fractions to concentrator tube during reflux
Supplier: Synthware™ F984250
Description
- Top joint: 24/40 outer
- Lower joint: 19/22 inner
Specifications
250 mL | |
Kuderna-danish Flask |