Kemtech America™ Synthware™ Gas Dispersion Tubes, Fritted
For use with filtration techniques
$42.97 - $44.06
Specifications
Description Gas dispersion tube, fritted
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Item Description Price Quantity  
Catalog Number Mfr. No. Item Description Price Quantity  
31501543 Kemtech America™
F260810X
Porosity: Extra Coarse Each for $42.97
31501544 Kemtech America™
F260810C
Porosity: Coarse Each for $42.97
31501545 Kemtech America™
F260810M
Porosity: Medium Each for $44.06
Specifications
Description
  • With stem O.D. of 8mm and Length: 150mm
  • Disc diameter: 10mm
Description & Specifications
Specifications
Gas dispersion tube, fritted