Kemtech America™ Synthware™ Flow Control Adapters with PTFE Stopcock and Outer Joint
For regulating the flow of gases or liquids into reaction systems
$86.31 - $92.86
Specifications
Product Type Flow control adapter
Includes PTFE Stopcock
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
31500560 Kemtech America™
A711420
14/20 Each for $86.31
31500561 Kemtech America™
A711922
19/22 Each for $87.26
31500562 Kemtech America™
A712440
24/40 Each for $92.86
Specifications
Description
  • Stopcock has bore of 2mm
  • With standard taper inner joint
Description & Specifications
Specifications
Flow control adapter
PTFE Stopcock