missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Flow Control Adapters with Glass Stopcock, 90°
To regulate the flow of gases or liquids into reaction systems
Supplier: Synthware™ A752440
Description
- Stopcock has a bore of 2mm
- With standard taper inner joint
Specifications
Borosilicate Glass (3.3) | |
Flow control adapter with glass stopcock | |
24/40 |