Synthware™ Filter Tubes with Fritted Disc
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Supplier: Synthware™ F701430M
Description
- With standard taper inner joints
- Available with either coarse or medium porosity fritted disc
Specifications
Approximate Frit O.D.: 30 mm, Medium porosity | |
Medium | |
Approximate Frit O.D.: 30 mm |
Borosilicate Glass (3.3) | |
14/20 |