Synthware™ Evaporation Flask, 14/20 Joint
For controlled evaporation of foaming solvents such as toluene or xylene on rotary evaporator
Supplier: Synthware™ F794200
Description
- Low profile design sits upright on bench
- Provides easy product access with spatula
Specifications
Borosilicate Glass (3.3) | |
Rotary Evaporator | |
24/40 |
Evaporation Flask | |
Foaming Solvents | |
200 mL |