Synthware™ Distilling Head, Short Path, Vigreux
Has a close coupled condenser and vacuum connection to offer an extremely short condensate travel path
$311.88 - $320.29
Description
- Thermometer stem and bulb serves as column packing
- Lower hose connection is for vacuum, upper hose connections are for cooling lines
- Used with 25 mm immersion thermometer
- Vigreux indentations improve efficiency
Specifications
Thermometer Joint: 10/18 |