Synthware™ Distilling Head, Short Path, Vigreux
Has a close coupled condenser and vacuum connection to offer an extremely short condensate travel path
Supplier:  Synthware™ D251014
 View more versions of this product
Catalog No. 31501019
Description
Description
  • Thermometer stem and bulb serves as column packing
  • Lower hose connection is for vacuum, upper hose connections are for cooling lines
  • Used with 25 mm immersion thermometer
  • Vigreux indentations improve efficiency
Specifications
Specifications
Thermometer Joint: 10/18
14/20