Kemtech America™ Synthware™ Distillation Connecting Adapters, Angled at 75°
For use as a component in atmospheric or vacuum distillations
$51.09 - $68.57
Specifications
Product Type Distillation Connecting Adapter
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
31500655 Kemtech America™
A351014
Joints: 10/18 (Top), 14/20 (Bottom), 14/20 (Side) Each for $53.54
31500656 Kemtech America™
A351019
Joints: 10/18 (Top), 19/22 (Bottom), 19/22 (Side) Each for $57.43
31500658 Kemtech America™
A351420
Joints: 14/20 (Top), 14/20 (Bottom), 14/20 (Side) Each for $51.09
31500657 Kemtech America™
A351024
Joints: 10/18 (Top), 24/40 (Bottom), 24/40 (Side) Each for $68.57
31500659 Kemtech America™
A351922
Joints: 19/22 (Top), 19/22 (Bottom), 19/22 (Side) Each for $54.89
31500660 Kemtech America™
A352440
Joints: 24/40 (Top), 24/40 (Bottom), 24/40 (Side) Each for $65.37
31500661 Kemtech America™
A351424
Joints: 14/20 (Top), 24/40 (Bottom), 24/40 (Side) Each for $64.60
31500662 Kemtech America™
A354914
Joints: 14/20 (Top), 24/40 (Bottom), 19/22 (Side) Each for $65.63
Specifications
Description
  • Standard taper outer joint at the top
  • Inner joints on bottom and side arm are at a 75° angle to one another
Description & Specifications
Specifications
Distillation Connecting Adapter