Synthware™ Distillation Adapters
With threaded thermometer port on the side arm
$129.59 - $173.85
Specifications
Includes | Open top cap and Viton™ O-ring |
---|---|
Material | Borosilicate Glass (3.3) |
Product Type | Distillation adapter |
Description
- Accommodates 6 to 7.7mm O.D. plain stem thermometer
- Sidearm with standard taper inner joint with drip tip
- Vertical tube has top outer and lower inner joint
- Supplied with open top cap and Viton™ O-ring
Specifications
Open top cap and Viton™ O-ring | |
Borosilicate Glass (3.3) | |
Distillation adapter |