Synthware™ Chromatography Reservoir, with Standard Taper Joints
With standard taper joints
Supplier: Synthware™ C182402
Description
- Used for flash chromatography
Specifications
250 mL | |
Top Outer Joint: 24/40, Bottom inner Joint: 24/40 |