Synthware™ Chromatography Column with Standard Taper Joint, Reservoir, PTFE Stopcock and Fritted Disc
For flash chromatography
Supplier: Synthware™ C393240C
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
- With coarse porosity sealed in fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
26 mm | |
24/40 | |
457 mm |
500 mL | |
2 mm bore |