Synthware™ Chromatography Column with PTFE Stopcock, Fritted Disc
Chromatography Column with PTFE Stopcock and Fritted Disc for flash chromatography.
Supplier: Synthware™ C361720C
Description
- General purpose chromatography column with PTFE stopcock.
- With reinforced beaded top and coarse porosity fritted disc to support column packing.
- Heavy-wall design increases safety.
Specifications
13.4 mm | |
2 mm bore | |
203 mm |
Chromatography Column | |
40 to 80 μm | |
17 mm |