Synthware™ Buchner Funnels with Inner Joint and Perforated Glass Plate
With sealed in perforated glass disc to support filter media
$153.60 - $405.17
Specifications
Product Type Buchner Funnels with Inner Joint and Perforated Glass Plate
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Joint Number Diameter (Metric) Disc Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Joint Number Diameter (Metric) Disc Price Quantity  
31501454
View Documents Promotion Details
Synthware™
F963014
30 mL 14/20 32mm
Each for $153.60
31501456
View Documents Promotion Details
Synthware™
F964014
60 mL 14/20 42mm
Each for $157.40
31501455
View Documents Promotion Details
Synthware™
F963024
30 mL 24/40 32mm
Each for $191.14
31501457
View Documents Promotion Details
Synthware™
F964024
60 mL 24/40 42mm
Each for $199.31
31501458
View Documents Promotion Details
Synthware™
F966024
150 mL 24/40 60mm
Each for $290.29
31501459
View Documents Promotion Details
Synthware™
F969024
600 mL 24/40 90mm
Each for $405.17
Description
Description
  • Lower standard taper inner joint
  • Hose connector for connection to vacuum source
Specifications
Specifications
Buchner Funnels with Inner Joint and Perforated Glass Plate