Synthware™ Buchner Filter Funnels
Used to filter particulates from a variety of substances
$46.54 - $1,627.14
Catalog Number | Mfr. No. | Capacity (Metric) | Porosity | Diameter (Metric) Disc | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Porosity | Diameter (Metric) Disc | Price | Quantity | |||
31501514
|
Synthware™
F601002M |
2 mL | Medium | 10mm |
Each for $46.54
|
|
31501515
|
Synthware™
F601002F |
2 mL | Fine | 10mm |
Each for $49.91
|
|
31501513
|
Synthware™
F601002C |
2 mL | Coarse | 10mm |
Each for $46.54
|
|
31501516
|
Synthware™
F602015C |
15 mL | Coarse | 20mm |
Each for $52.29
|
|
31501517
|
Synthware™
F602015X |
15 mL | Extra Coarse | 20mm |
Each for $52.29
|
|
31501518
|
Synthware™
F602015M |
15 mL | Medium | 20mm |
Each for $52.29
|
|
31501519
|
Synthware™
F602015F |
15 mL | Fine | 20mm |
Each for $55.57
|
|
31501521
|
Synthware™
F603030C |
30 mL | Coarse | 30mm |
Each for $58.89
|
|
31501520
|
Synthware™
F603030X |
30 mL | Extra Coarse | 30mm |
Each for $58.89
|
|
31501523
|
Synthware™
F603030F |
30 mL | Fine | 30mm |
Each for $62.17
|
|
31501522
|
Synthware™
F603030M |
30 mL | Medium | 30mm |
Each for $58.89
|
|
31501525
|
Synthware™
F604060C |
60 mL | Coarse | 40mm |
Each for $65.00
|
|
31501526
|
Synthware™
F604060M |
60 mL | Medium | 40mm |
Each for $65.00
|
|
31501524
|
Synthware™
F604060X |
60 mL | Extra Coarse | 40mm |
Each for $65.00
|
|
31501527
|
Synthware™
F604060F |
60 mL | Fine | 40mm |
Each for $68.29
|
|
31501528
|
Synthware™
F606150X |
150 mL | Extra Coarse | 60mm |
Each for $115.94
|
|
31501529
|
Synthware™
F606150C |
150 mL | Coarse | 60mm |
Each for $115.94
|
|
31501530
|
Synthware™
F606150M |
150 mL | Medium | 60mm |
Each for $115.94
|
|
31501531
|
Synthware™
F606150F |
150 mL | Fine | 60mm |
Each for $119.17
|
|
31501532
|
Synthware™
F608350X |
350 mL | Extra Coarse | 80mm |
Each for $160.91
|
|
31501533
|
Synthware™
F608350C |
350 mL | Coarse | 80mm |
Each for $160.91
|
|
31501534
|
Synthware™
F608350M |
350 mL | Medium | 80mm |
Each for $160.91
|
|
31501535
|
Synthware™
F608350F |
350 mL | Fine | 80mm |
Each for $164.29
|
|
31501537
|
Synthware™
F609600M |
600 mL | Medium | 90mm |
Each for $192.94
|
|
31501536
|
Synthware™
F609600C |
600 mL | Coarse | 90mm |
Each for $192.94
|
|
31501538
|
Synthware™
F609600F |
600 mL | Fine | 90mm |
Each for $196.31
|
|
31-502-161
|
Synthware™
F601000X |
1000 mL | Extra Course | 120 mm |
Each for $385.37
|
|
31-502-160
|
Synthware™
F601000C |
1000 mL | Course | 120 mm |
Each for $385.37
|
|
31-502-159
|
Synthware™
F601000M |
1000 mL | Medium | 120 mm |
Each for $385.37
|
|
31-502-158
|
Synthware™
F601000F |
1000 mL | Fine | 120 mm |
Each for $419.63
|
|
31-502-157
|
Synthware™
F602000X |
2000 mL | Extra Course | 150 mm |
Each for $770.74
|
|
31-502-156
|
Synthware™
F602000C |
2000 mL | Course | 150 mm |
Each for $770.74
|
|
31-502-155
|
Synthware™
F602000M |
2000 mL | Medium | 150 mm |
Each for $770.74
|
|
31-502-154
|
Synthware™
F602000F |
2000 mL | Fine | 150 mm |
Each for $839.26
|
|
31-502-153
|
Synthware™
F605000X |
5000 mL | Extra Course | 180 mm |
Each for $1,541.49
|
|
31-502-152
|
Synthware™
F605000C |
5000 mL | Course | 180 mm |
Each for $1,541.49
|
|
31-502-151
|
Synthware™
F605000M |
5000 mL | Medium | 180 mm |
Each for $1,541.49
|
|
31-502-150
|
Synthware™
F605000F |
5000 mL | Fine | 180 mm |
Each for $1,627.14
|
|
Description
- Sealed in glass fritted disc of listed size and porosity
- Porosity: XC-Extra Coarse, C-Coarse, M-Medium, F-Fine
- Not recommended for filtration where pressure would exceed 15 psi
Specifications
Buchner filter funnel |